2-Methylcyclohexylamine (cis- and trans- mixture) 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L02285-09
L02285-18
CAAAAL02285-09EA
53.25
CAD
CAAAAL02285-09
CAAAAL02285-18
2-Methylcyclohexylamine (cis- and trans- mixture) 97%
2-Methylcyclohexylamine (cis- and trans- mixture)
Purity: 97%
CAS No.: 7003-32-9
Molecular Formula: C7H15N
Formula Weight: 113.20
Flash Point: 21°(69°F)
Refractive Index: 1.4565
Boiling Point: 149-150°
Beilstein Reference: 2689073
UN No.: 2733
MDL No.: MFCD00001492
Formula:
C₇H₁₅N Boiling Pt: 149…150 °C Density: 0.858 Flash Pt: 21 °C(69 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001492 CAS Number: 7003-32-9 EINECS: 230-277-5 UN: 2733 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...