Cyclopropyl phenyl ketone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzoylcyclopropane, Cyclopropyl(phenyl)methanone

L02218-14 L02218-22
CAAAAL02218-14EA 120.71 CAD
CAAAAL02218-14 CAAAAL02218-22
Cyclopropyl phenyl ketone 97%
Cyclopropyl phenyl ketone

Purity: 97% CAS No.: 3481-02-5 Molecular Formula: C10H10O Formula Weight: 146.19 Flash Point: 90°(194°F) Refractive Index: 1.5534 Boiling Point: 248° Beilstein Reference: 1860145 MDL No.: MFCD00001294

Formula: C₁₀H₁₀O
MW: 146.19 g/mol
Boiling Pt: 248 °C
Density: 1.06
Flash Pt: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001294
CAS Number: 3481-02-5
EINECS: 222-458-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR