1-Methyl-1H-indole-3-carbaldehyde ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Methylindole-3-carboxaldehyde
L02212-03
L02212-06
CAAAAL02212-03EA
57.99
CAD
CAAAAL02212-03
CAAAAL02212-06
1-Methyl-1H-indole-3-carbaldehyde ≥98%
1-Methyl-1H-indole-3-carbaldehyde
Purity: 98+%
CAS No.: 19012-03-4
Molecular Formula: C10H9NO
Formula Weight: 159.19
Melting Point: 67-71°
Beilstein Reference: 121302
MDL No.: MFCD00014570
Formula:
C₁₀H₉NO Melting Pt: 67…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014570 CAS Number: 19012-03-4 EINECS: 242-750-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...