N-Methylhydroxylammonium chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Methylhydroxylamine hydrochloride

L02202-06 L02202-14
CAAAAL02202-06EA 111.07 CAD
CAAAAL02202-06 CAAAAL02202-14
N-Methylhydroxylammonium chloride 98%
N-Methylhydroxylammonium chloride

Fieser: 7,241 12,322


Purity: 98% CAS No.: 4229-44-1 Molecular Formula: CH3NHOH.HCl Formula Weight: 83.52 Melting Point: 80-86° Beilstein Reference: 3541409

Formula: CH₆ClNO
MW: 83.52 g/mol
Melting Pt: 80…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00012597
CAS Number: 4229-44-1
EINECS: 224-181-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR