Diphenyl phosphate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Diphenyl hydrogen phosphate

L02121-06 L02121-14 L02121-22
CAAAAL02121-06EA 65.25 CAD
CAAAAL02121-06 CAAAAL02121-14 CAAAAL02121-22
Diphenyl phosphate 97%
Diphenyl phosphate

Purity: 97% CAS No.: 838-85-7 Molecular Formula: C12H11O4P Formula Weight: 250.19 Melting Point: 62-70° Beilstein Reference: 1379164

Formula: C₁₂H₁₁O₄P
MW: 250.19 g/mol
Melting Pt: 62…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00003033
CAS Number: 838-85-7
EINECS: 212-657-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR