2-(p-Toluenesulfonyl)acetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L02102-06
L02102-14
CAAAAL02102-06EA
150.1
CAD
CAAAAL02102-06
CAAAAL02102-14
2-(p-Toluenesulfonyl)acetic acid 98%
2-(p-Toluenesulfonyl)acetic acid
Purity: 98%
CAS No.: 3937-96-0
Molecular Formula: C9H10O4S
Formula Weight: 214.24
Melting Point: 117-119°
Beilstein Reference: 2331311
MDL No.: MFCD00021764
Formula:
C₉H₁₀O₄S Melting Pt: 117…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021764 CAS Number: 3937-96-0 EINECS: 223-518-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...