5,6-Dihydro-5-methyluracil ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: 5-Methyldihydropyrimidine-2,4(1H,3H)-dione

L01996-06 L01996-14
CAAAAL01996-06EA 61.62 CAD
CAAAAL01996-06 CAAAAL01996-14
5,6-Dihydro-5-methyluracil ≥98%
5,6-Dihydro-5-methyluracil

Purity: 98+% CAS No.: 696-04-8 Molecular Formula: C5H8N2O2 Formula Weight: 128.13 Melting Point: 263-265° Beilstein Reference: 81983 MDL No.: MFCD00023159

Formula: C₅H₈N₂O₂
MW: 128.13 g/mol
Melting Pt: 263…265 °C
Storage Temperature: Ambient
MDL Number: MFCD00023159
CAS Number: 696-04-8
EINECS: 211-787-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR