5-Hydroxymethyluracil 98%

Supplier: Thermo Scientific Chemicals

L01682-03 L01682-06
CAAAAL01682-03EA 66.52 CAD
CAAAAL01682-03 CAAAAL01682-06
5-Hydroxymethyluracil 98%
5-Hydroxymethyluracil

Purity: 98% CAS No.: 4433-40-3 Molecular Formula: C5H6N2O3 Formula Weight: 142.12 Melting Point: 300° Beilstein Reference: 125482 MDL No.: MFCD00006070

Formula: C₅H₆N₂O₃
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006070
CAS Number: 4433-40-3
EINECS: 224-636-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR