2-Acetamidoacrylic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L01649-06
L01649-14
CAAAAL01649-03EA
125.4
CAD
CAAAAL01649-03
CAAAAL01649-06
CAAAAL01649-14
2-Acetamidoacrylic acid 99%
2-Acetamidoacrylic acid
Fieser: 15,53
Purity: 99%
CAS No.: 5429-56-1
Molecular Formula: C5H7NO3
Formula Weight: 129.12
Storage Temperature: 2°C to 8°C
Melting Point: ca 190° dec.
Beilstein Reference: 1812032
MDL No.: MFCD00004257
Formula:
C₅H₇NO₃ Melting Pt: ∼ 190 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004257 CAS Number: 5429-56-1 EINECS: 226-583-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...