D(-)-2-Phenylglycine ethyl ester hydrochloride ≥98%
Supplier: Thermo Scientific Chemicals
L01564-09
L01564-18
CAAAAL01564-09EA
145.68
CAD
CAAAAL01564-09
CAAAAL01564-18
D(-)-2-Phenylglycine ethyl ester hydrochloride ≥98%
D(-)-2-Phenylglycine ethyl ester hydrochloride
Purity: 98+%
CAS No.: 17609-48-2
Molecular Formula: C10H13NO2.HCl
Formula Weight: 215.68
Melting Point: ca 190° dec.
Beilstein Reference: 5775455
Optical Rotation: -94° (c=1 in water)
MDL No.: MFCD00052005
Formula:
C₁₀H₁₃NO₂·HCl Melting Pt: ca 190 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00052005 CAS Number: 17609-48-2 EINECS: 241-581-2 |
Specification Test Results
Optical rotation | -94° (c=1 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...