Methyl vinyl sulfone 95% stabilized

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: 200 ppm Stabilised with 4-tert-butylphenol

L01519-03 L01519-06
CAAAAL01519-03EA 48.19 CAD
CAAAAL01519-03 CAAAAL01519-06
Methyl vinyl sulfone 95% stabilized
Methyl vinyl sulphone

Purity: 95% CAS No.: 3680-02-2 Molecular Formula: C3H6O2S Formula Weight: 106.15 Storage Temperature: 2°C to 8°C Flash Point: 109°(228°F) Refractive Index: 1.4630 Boiling Point: 115-117°/19mm Beilstein Reference: 878198 UN No.: 2810 MDL No.: MFCD00007567

Formula: C₃H₆O₂S
MW: 106.15 g/mol
Boiling Pt: 115…117 °C (19 mmHg)
Density: 1.212
Flash Pt: >109 °C (228 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007567
CAS Number: 3680-02-2
EINECS: 222-958-0
UN: 2810
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR