5-Nitrothiophene-2-carboxaldehyde 98%

Supplier: Thermo Scientific Chemicals

L01493-06 L01493-14
CAAAAL01493-06EA 152.47 CAD
CAAAAL01493-06 CAAAAL01493-14
5-Nitrothiophene-2-carboxaldehyde 98%
5-Nitrothiophene-2-carboxaldehyde

Purity: 98% CAS No.: 4521-33-9 Molecular Formula: C5H3NO3S Formula Weight: 157.15 Storage Temperature: 2°C to 8°C Melting Point: 72-76° Beilstein Reference: 120540 MDL No.: MFCD00005433

Formula: C5H3NO3S
MW: 157.15 g/mol
Melting Pt: 72…76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005433
CAS Number: 4521-33-9
EINECS: 224-850-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR