3-Fluoro-DL-tyrosine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-DL-Tyr(3-F)-OH, 2-Amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid

L01479-MC L01479-ME
CAAAAL01479-MCEA 133.98 CAD
CAAAAL01479-MC CAAAAL01479-ME
3-Fluoro-DL-tyrosine 97%
3-Fluoro-DL-tyrosine

Purity: 98% CAS No.: 403-90-7 Molecular Formula: C9H10FNO3 Formula Weight: 199.18 Melting Point: ca 280° dec. Beilstein Reference: 3204804 MDL No.: MFCD00063075

Formula: C₉H₁₀FNO₃
Melting Pt: ∼ 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00063075
CAS Number: 403-90-7
EINECS: 206-964-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR