2-Carboxycinnamic acid predominantly trans 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L01467-06
L01467-14
CAAAAL01467-06EA
80.26
CAD
CAAAAL01467-06
CAAAAL01467-14
2-Carboxycinnamic acid predominantly trans 97%
2-Carboxycinnamic acid
Purity: 97%
CAS No.: 612-40-8
Molecular Formula: C10H8O4
Formula Weight: 192.17
Melting Point: 195-205°
Beilstein Reference: 1870294
MDL No.: MFCD00004380
Formula:
C₁₀H₈O₄ Melting Pt: 195…205 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004380 CAS Number: 612-40-8 EINECS: 210-308-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...