Cinnamyl bromide predominantly trans 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(3-Bromo-1-propen-1-yl)benzene, 3-Bromo-1-phenyl-1-propene
L01420-06
L01420-14
L01420-22
CAAAAL01420-06EA
58.3
CAD
CAAAAL01420-06
CAAAAL01420-14
CAAAAL01420-22
Cinnamyl bromide predominantly trans 95%
Cinnamyl bromide
Purity: 95%
CAS No.: 4392-24-9
Molecular Formula: C6H5CH=CHCH2Br
Formula Weight: 197.08
Flash Point: 87°(188°F)
Refractive Index: 1.6210
Melting Point: 26-29°
Boiling Point: 88-90°/1mm
Beilstein Reference: 605833
UN No.: 3261
MDL No.: MFCD00000245
Formula:
C₉H₉Br MW: 197.08 g/mol Boiling Pt: 88…90 °C/1 mm Melting Pt: 26…29 °C Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000245 CAS Number: 4392-24-9 EINECS: 224-511-5 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...