2-(2-Pyridyl)benzimidazole ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: AI-4-88

L01339-03 L01339-06
CAAAAL01339-03EA 38.08 CAD
CAAAAL01339-03 CAAAAL01339-06
2-(2-Pyridyl)benzimidazole ≥98%
2-(2-Pyridyl)benzimidazole

Purity: 98+% CAS No.: 1137-68-4 Molecular Formula: C12H9N3 Formula Weight: 195.22 Melting Point: 221-225° Beilstein Reference: 151411 MDL No.: MFCD00005586

Formula: C₁₂H₉N₃
MW: 195.23 g/mol
Melting Pt: 221…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00005586
CAS Number: 1137-68-4
EINECS: 214-508-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR