2-Bromoacrylic acid 95%

Supplier: Thermo Scientific Chemicals
Danger

L01194-06 L01194-14
CAAAAL01194-06EA 110.13 CAD
CAAAAL01194-06 AAL01194-14
2-Bromoacrylic acid 95%
2-Bromoacrylic acid

Purity: 95% CAS No.: 10443-65-9 Molecular Formula: C3H3BrO2 Formula Weight: 150.96 Storage Temperature: 2°C to 8°C Melting Point: 65-70° Beilstein Reference: 1740582 UN No.: 3261 MDL No.: MFCD00014333

Formula: C₃H₃BrO₂
Melting Pt: 65…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014333
CAS Number: 10443-65-9
EINECS: 233-928-1
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR