(Cyclopropylmethyl)triphenylphosphonium bromide 98+%

Supplier: Thermo Scientific Chemicals
Warning

L01145-06 L01145-14
CAAAAL01145-06EA 164.32 CAD
CAAAAL01145-06 CAAAAL01145-14
(Cyclopropylmethyl)triphenylphosphonium bromide 98+%
(Cyclopropylmethyl)triphenylphosphonium bromide

Purity: 98+% CAS No.: 14799-82-7 Molecular Formula: C22H22BrP Formula Weight: 397.29 Melting Point: 178-182° Beilstein Reference: 4068302

Formula: C₁₄H₂₉ClNP
Melting Pt: 178…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00051878
CAS Number: 14799-82-7
EINECS: 238-862-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR