5-Benzyloxyindole-3-carbaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-(Benzyloxy)-1H-indole-3-carbaldehyde, 5-benzyloxy-1h-indole-3-carbaldehyde, 5-Benzyloxyindole-3-carboxaldehyde

L01117-03 L01117-06
CAAAAL01117-03EA 102.38 CAD
CAAAAL01117-03 CAAAAL01117-06 CAAAAL01117-MD
5-Benzyloxyindole-3-carbaldehyde 98%
5-Benzyloxyindole-3-carbaldehyde

Purity: 98% CAS No.: 6953-22-6 Molecular Formula: C16H13NO2 Formula Weight: 251.28 Melting Point: 239-242° Beilstein Reference: 208873 MDL No.: MFCD00014562

Formula: C₁₆H₁₃NO₂
MW: 251.285 g/mol
Melting Pt: 239…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00014562
CAS Number: 6953-22-6
EINECS: 230-134-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR