2-(4-Isopropylphenoxy)acetic acid ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L01101-06
L01101-14
CAAAAL01101-06EA
77.1
CAD
CAAAAL01101-06
CAAAAL01101-14
2-(4-Isopropylphenoxy)acetic acid ≥98%
2-(4-Isopropylphenoxy)acetic acid
Purity: 98+%
CAS No.: 1643-16-9
Molecular Formula: C11H14O3
Formula Weight: 194.23
Melting Point: 85-87°
Beilstein Reference: 2261017
MDL No.: MFCD00014364
Formula:
C₁₁H₁₄O₃ Melting Pt: 85…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014364 CAS Number: 1643-16-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...