4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl 96% free radical
Supplier: Thermo Scientific Chemicals
L01084-03
L01084-06
CAAAAL01084-03EA
82.95
CAD
CAAAAL01084-03
CAAAAL01084-06
4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl 96% free radical
4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl
Purity: 96%
CAS No.: 2896-70-0
Molecular Formula: C9H16NO2
Formula Weight: 170.23
Melting Point: 36-40°
Beilstein Reference: 1818579
MDL No.: MFCD00006187
Formula:
C₉H₁₆NO₂ Melting Pt: 36…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006187 CAS Number: 2896-70-0 EINECS: 220-778-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...