2-(8-Bromooctyloxy)tetrahydropyran 90%, tech.

Supplier: Thermo Scientific Chemicals

L01081-06 L01081-14
CAAAAL01081-03EA 79.9 CAD
CAAAAL01081-03 CAAAAL01081-06 CAAAAL01081-14
2-(8-Bromooctyloxy)tetrahydropyran 90%, tech.
2-(8-Bromooctyloxy)tetrahydropyran

Purity: 90% CAS No.: 50816-20-1 Molecular Formula: C13H25BrO2 Formula Weight: 293.25 Flash Point: 110°(230°F) Refractive Index: 1.4770 Boiling Point: 130-132°/0.8mm Beilstein Reference: 1306644 MDL No.: MFCD00014648

Formula: C₁₃H₂₅BrO₂
Boiling Pt: 130…132 °C (0.8 mmHg)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014648
CAS Number: 50816-20-1
EINECS: 256-786-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR