5-Phenyl-2,4-pentadienoic acid ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L01004-06
L01004-14
CAAAAL01004-06EA
146.31
CAD
CAAAAL01004-06
CAAAAL01004-14
5-Phenyl-2,4-pentadienoic acid ≥98%
5-Phenyl-2,4-pentadienoic acid
Purity: 98+%
CAS No.: 1552-94-9
Molecular Formula: C6H5CH=CHCH=CHCHCO2H
Formula Weight: 174.20
Melting Point: 163-167°
Beilstein Reference: 2042414
MDL No.: MFCD00014018
Formula:
C₆H₅CH=CHCH=CHCHCO₂H Melting Pt: 163…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014018 CAS Number: 1552-94-9 EINECS: 216-298-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...