Diethyl (4-chlorobenzyl)phosphonate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diethyl 4-chlorobenzylphosphonate
L00965-09
L00965-18
CAAAAL00965-09EA
89.43
CAD
CAAAAL00965-09
CAAAAL00965-18
Diethyl (4-chlorobenzyl)phosphonate 98+%
Diethyl (4-chlorobenzyl)phosphonate
Purity: 98+%
CAS No.: 39225-17-7
Molecular Formula: C11H16ClO3P
Formula Weight: 262.67
Flash Point:
Formula:
C₁₁H₁₆ClO₃P MW: 262.67 g/mol Boiling Pt: 153…155 °C/0.6 mm Density: 1,082 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051568 CAS Number: 39225-17-7 EINECS: 254-365-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...