2-(2-Bromoethyl)-1,3-dioxane 98%

Supplier: Thermo Scientific Chemicals
Warning

L00958-09 L00958-18
CAAAAL00958-09EA 64.15 CAD
CAAAAL00958-09 CAAAAL00958-18
2-(2-Bromoethyl)-1,3-dioxane 98%
2-(2-Bromoethyl)-1,3-dioxane

Fieser: 7,37 11,78


Purity: 98% CAS No.: 33884-43-4 Molecular Formula: C6H11BrO2 Formula Weight: 195.06 Storage Temperature: 2°C to 8°C Flash Point: 96°(204°F) Refractive Index: 1.4820 Boiling Point: 67-70°/3mm Beilstein Reference: 1421628 MDL No.: MFCD00006567

Formula: C₆H₁₁BrO₂
MW: 195.06 g/mol
Boiling Pt: 67…70 °C (3 mmHg)
Density: 1.434
Flash Pt: 96 °C (204 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00006567
CAS Number: 33884-43-4
EINECS: 251-716-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR