Dibenzyl ketoxime ≥98%

Supplier: Thermo Scientific Chemicals

L00914-06 L00914-14
CAAAAL00914-06EA 55.3 CAD
CAAAAL00914-06 CAAAAL00914-14
Dibenzyl ketoxime ≥98%
Dibenzyl ketoxime

Purity: 98+% CAS No.: 1788-31-4 Molecular Formula: C15H15NO Formula Weight: 225.29 Melting Point: 122-124° Beilstein Reference: 1965977 MDL No.: MFCD00015453

Formula: C₁₅H₁₅NO
MW: 225.29 g/mol
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00015453
CAS Number: 1788-31-4
EINECS: 217-251-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR