(Methoxymethyl)diphenylphosphine oxide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00912-06
L00912-14
CAAAAL00912-06EA
134.93
CAD
CAAAAL00912-06
CAAAAL00912-14
(Methoxymethyl)diphenylphosphine oxide 98+%
(Methoxymethyl)diphenylphosphine oxide
Purity: 98+%
CAS No.: 4455-77-0
Molecular Formula: C14H15O2P
Formula Weight: 246.24
Melting Point: 113-120°
Beilstein Reference: 649558
Formula:
C₁₄H₁₅O₂P MW: 246.24 g/mol Melting Pt: 113…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015451 CAS Number: 4455-77-0 EINECS: 224-704-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...