2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate, tech.

Supplier: Thermo Scientific Chemicals
Warning

L00875-03 L00875-06
CAAAAL00875-03EA 155.16 CAD
CAAAAL00875-03 CAAAAL00875-06 CAAAAL00875-14
2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate, tech.
2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate

CAS No.: 375-02-0 Molecular Formula: CF3CF2CF3CHO.xH2O Formula Weight: 198.04(anhy) Refractive Index: 1.3200 Beilstein Reference: 1367365 MDL No.: MFCD00150588

Formula: CF₃CF₂CF₂CHO·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00150588
CAS Number: 375-02-0
EINECS: 206-783-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR