3-Bromo-2,4,6-trimethylaniline ≥98%

Supplier: Thermo Scientific Chemicals
Warning

L00873-09 L00873-18
CAAAAL00873-09EA 119.45 CAD
CAAAAL00873-09 CAAAAL00873-18
3-Bromo-2,4,6-trimethylaniline ≥98%
3-Bromo-2,4,6-trimethylaniline

Purity: 98+% CAS No.: 82842-52-2 Molecular Formula: C9H12BrN Formula Weight: 214.11 Flash Point: 110°(230°F) Melting Point: 34-38° Boiling Point: 153-155°/17mm Beilstein Reference: 2829829 MDL No.: MFCD00015476

Formula: C₉H₁₂BrN
Boiling Pt: 153…155 °C/17 mm
Melting Pt: 34…38 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015476
CAS Number: 82842-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR