Phenylpropiolaldehyde diethyl acetal 98%
Supplier: Thermo Scientific Chemicals
L00872-06
L00872-14
CAAAAL00872-06EA
50.88
CAD
CAAAAL00872-06
CAAAAL00872-14
Phenylpropiolaldehyde diethyl acetal 98%
Phenylpropiolaldehyde diethyl acetal
Purity: 98%
CAS No.: 6142-95-6
Molecular Formula: C6H5C≡CCH(OCH2CH3)2
Formula Weight: 204.27
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₆H₅CΞCCH(OCH₂CH₃)₂ Boiling Pt: 99…100 °C/2 mm Density: 0.991 Flash Pt: >110 °C(230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009238 CAS Number: 6142-95-6 EINECS: 228-143-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...