α-Methylcinnamic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00806-06
L00806-14
L00806-22
CAAAAL00806-06EA
57.83
CAD
CAAAAL00806-06
CAAAAL00806-14
CAAAAL00806-22
α-Methylcinnamic acid 99%
α-Methylcinnamic acid
Purity: 99%
CAS No.: 1199-77-5
Molecular Formula: C10H10O2
Formula Weight: 162.19
Melting Point: 78-82°
Boiling Point: 288°
MDL No.: MFCD00002652
Formula:
C₁₀H₁₀O₂ Boiling Pt: 288 °C Melting Pt: 78…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002652 CAS Number: 1199-77-5 EINECS: 214-847-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...