2-Chloro-2,2-difluoroacetamide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00785-06
L00785-14
CAAAAL00785-06EA
251.22
CAD
CAAAAL00785-06
CAAAAL00785-14
2-Chloro-2,2-difluoroacetamide 98%
2-Chloro-2,2-difluoroacetamide
Purity: 98%
CAS No.: 354-28-9
Molecular Formula: ClCF2CONH2
Formula Weight: 129.49
Melting Point: 79-84°
Beilstein Reference: 1753808
MDL No.: MFCD00039775
Formula:
ClCF₂CONH₂ MW: 129.49 g/mol Melting Pt: 79…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039775 CAS Number: 354-28-9 EINECS: 206-555-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...