Ethyl-5-hydroxyindole-2-carboxylate ≥98%
Supplier: Thermo Scientific Chemicals
L00774-03
L00774-06
CAAAAL00774-03EA
133.67
CAD
CAAAAL00774-03
CAAAAL00774-06
CAAAAL00774-MD
Ethyl-5-hydroxyindole-2-carboxylate ≥98%
Ethyl-5-hydroxyindole-2-carboxylate
Purity: 98+%
CAS No.: 24985-85-1
Molecular Formula: C11H11NO3
Formula Weight: 205.21
Storage Temperature: 2°C to 8°C
Melting Point: 150-151°
Beilstein Reference: 476925
MDL No.: MFCD00015458
Formula:
C₁₁H₁₁NO₃ Melting Pt: 150…151 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00015458 CAS Number: 24985-85-1 EINECS: 246-554-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...