Methyl cyclopentanecarboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00741-06
L00741-14
CAAAAL00741-06EA
139.2
CAD
CAAAAL00741-06
CAAAAL00741-14
Methyl cyclopentanecarboxylate 97%
Methyl cyclopentanecarboxylate
Purity: 97%
CAS No.: 4630-80-2
Molecular Formula: C7H12O2
Formula Weight: 128.17
Refractive Index: 1.4360
Boiling Point: 81-82°/5mm
MDL No.: MFCD00019290
Formula:
C₇H₁₂O₂ Boiling Pt: 81…82 °C/5 mm Storage Temperature: Ambient |
MDL Number:
MFCD00019290 CAS Number: 4630-80-2 EINECS: 225-049-7 UN: 3272 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...