5,6-Dimethoxy-1H-indole-2-carboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00733-03
L00733-06
CAAAAL00733-03EA
164.32
CAD
CAAAAL00733-03
CAAAAL00733-06
5,6-Dimethoxy-1H-indole-2-carboxylic acid 98%
5,6-Dimethoxy-1H-indole-2-carboxylic acid
Purity: 98%
CAS No.: 88210-96-2
Molecular Formula: C11H11NO4
Formula Weight: 221.22
Melting Point: ca 210° dec.
Beilstein Reference: 202249
MDL No.: MFCD00015459
Formula:
C₁₁H₁₁NO₄ Melting Pt: ca 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00015459 CAS Number: 88210-96-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...