2-Chloro-6-methylbenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

L00706-03 L00706-06
CAAAAL00706-03EA 52.14 CAD
CAAAAL00706-03 CAAAAL00706-06
2-Chloro-6-methylbenzonitrile 98%
2-Chloro-6-methylbenzonitrile

Purity: 98% CAS No.: 6575-09-3 Molecular Formula: C8H6ClN Formula Weight: 151.60 Odor: Characteristic Melting Point: 79-83° Boiling Point: 143°/38mm Beilstein Reference: 2246414 UN No.: 3439 MDL No.: MFCD00001782

Formula: C₈H₆ClN
Boiling Pt: 143 °C/38 mm
Melting Pt: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00001782
CAS Number: 6575-09-3
EINECS: 229-499-5
UN: 3439
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR