Cyclohexyldiphenylphosphine oxide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00698-03
L00698-06
CAAAAL00698-03EA
59.09
CAD
CAAAAL00698-03
CAAAAL00698-06
Cyclohexyldiphenylphosphine oxide 98+%
Cyclohexyldiphenylphosphine oxide
Purity: 98+%
CAS No.: 13689-20-8
Molecular Formula: C18H21OP
Formula Weight: 284.34
Melting Point: 166-167°
Beilstein Reference: 2943737
Formula:
C₁₈H₂₁OP Melting Pt: 166…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014301 CAS Number: 13689-20-8 EINECS: 237-204-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...