Phenylglyoxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00648-06
L00648-14
CAAAAL00648-06EA
66.2
CAD
CAAAAL00648-06
CAAAAL00648-14
Phenylglyoxylic acid 98%
Phenylglyoxylic acid
Purity: 98%
CAS No.: 611-73-4
Molecular Formula: C8H6O3
Formula Weight: 150.13
Melting Point: 62-67°
Boiling Point: 147-151°/12mm
MDL No.: MFCD00002575
Formula:
C₈H₆O₃ Boiling Pt: 147…151 °C (12 mmHg) Melting Pt: 62…67 °C Density: 1,38 Storage Temperature: Ambient |
MDL Number:
MFCD00002575 CAS Number: 611-73-4 EINECS: 210-278-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...