Trimethylsulfonium tetrafluoroborate ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L00647-06
L00647-14
CAAAAL00647-06EA
341.28
CAD
CAAAAL00647-06
CAAAAL00647-14
Trimethylsulfonium tetrafluoroborate ≥98%
Trimethylsulfonium tetrafluoroborate
Purity: 98+%
CAS No.: 676-88-0
Molecular Formula: (CH3)3SBF4
Formula Weight: 163.98
Storage Temperature: 2°C to 8°C
Melting Point: 214-218°
Beilstein Reference: 3708437
UN No.: 3261
MDL No.: MFCD00031582
Formula:
(CH₃)₃SBF₄ Melting Pt: 214…218 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00031582 CAS Number: 676-88-0 EINECS: 211-632-3 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...