S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine 98%
Supplier: Thermo Scientific Chemicals
L00577-06
L00577-14
CAAAAL00577-06EA
59.88
CAD
CAAAAL00577-06
CAAAAL00577-14
S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine 98%
S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine
Fieser: 3,205 9,193 12,216
Purity: 98%
CAS No.: 22236-45-9
Molecular Formula: C9H13NO3S2
Formula Weight: 247.34
Melting Point: 167-169°
Beilstein Reference: 2119329
MDL No.: MFCD00013922
Formula:
C₉H₁₃NO₃S₂ Melting Pt: 167…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013922 CAS Number: 22236-45-9 EINECS: 244-861-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...