Diethyl-2-bromo-2-methylmalonate 98%

Supplier: Thermo Scientific Chemicals
Danger

L00575-14 L00575-22
CAAAAL00575-14EA 172.22 CAD
CAAAAL00575-14 CAAAAL00575-22
Diethyl-2-bromo-2-methylmalonate 98%
Diethyl-2-bromo-2-methylmalonate

Purity: 98% CAS No.: 29263-94-3 Molecular Formula: C8H13BrO4 Formula Weight: 253.10 Flash Point: 110°(230°F) Refractive Index: 1.4490 Boiling Point: 105-106°/5mm Beilstein Reference: 1779673 UN No.: 3265 MDL No.: MFCD00009125

Formula: C₈H₁₃BrO₄
Boiling Pt: 105…106 °C/5 mm
Density: 1.323
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009125
CAS Number: 29263-94-3
EINECS: 249-541-6
UN: 3265
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR