Diethyl benzylidenemalonate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Diethyl Benzalmalonate, Benzalmalonic Acid Diethyl Ester, Benzylidenemalonic Acid Diethyl Ester

L00561-14 L00561-22
CAAAAL00561-14EA 79.63 CAD
CAAAAL00561-14 CAAAAL00561-22
Diethyl benzylidenemalonate 98%
Diethyl benzylidenemalonate

Purity: 98% CAS No.: 5292-53-5 Molecular Formula: C14H16O4 Formula Weight: 248.28 Flash Point: 110°(230°F) Refractive Index: 1.5360 Melting Point: 28-31° Boiling Point: 215-217°/30mm Beilstein Reference: 528894 MDL No.: MFCD00009149

Formula: C₁₄H₁₆O₄
Boiling Pt: 215…217 °C/30 mm
Melting Pt: 28…31 °C
Density: 1.107
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009149
CAS Number: 5292-53-5
EINECS: 226-137-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR