Dimethyl (2-oxopropyl)phosphonate 97%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Dimethyl acetylmethylphosphonate
L00537-06
L00537-14
CAAAAL00537-06EA
175.38
CAD
CAAAAL00537-06
CAAAAL00537-14
Dimethyl (2-oxopropyl)phosphonate 97%
Dimethyl (2-oxopropyl)phosphonate
Purity: 97%
CAS No.: 4202-14-6
Molecular Formula: C5H11O4P
Formula Weight: 166.12
Flash Point: 93°(199°F)
Refractive Index: 1.4390
Boiling Point: 121-123°/11mm
Beilstein Reference: 1766218
MDL No.: MFCD00008769
Formula:
C₅H₁₁O₄P MW: 166.12 g/mol Boiling Pt: 121…123 °C/11 mm Density: 1,202 Flash Pt: 93 °C(199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008769 CAS Number: 4202-14-6 EINECS: 224-110-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...