(2-Butenyl)triphenylphosphonium chloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00502-06
L00502-14
CAAAAL00502-06EA
90.22
CAD
CAAAAL00502-06
CAAAAL00502-14
(2-Butenyl)triphenylphosphonium chloride 97%
(2-Butenyl)triphenylphosphonium chloride
Purity: 97%
CAS No.: 13138-25-5
Molecular Formula: CH3CH=CHCH2P(C6H5)3Cl
Formula Weight: 352.84
Melting Point: ca 225-230° dec.
Beilstein Reference: 4170527
Formula:
C₂₂H₂₂ClP Melting Pt: ca 225…230 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00051868 CAS Number: 13138-25-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...