4-Methoxybenzhydrol 98+%

Supplier: Thermo Scientific Chemicals

L00468-06 L00468-14
CAAAAL00468-06EA 65.73 CAD
CAAAAL00468-06 CAAAAL00468-14
4-Methoxybenzhydrol 98+%
4-Methoxybenzhydrol

Purity: 98+% CAS No.: 720-44-5 Molecular Formula: C14H14O2 Formula Weight: 214.26 Storage Temperature: 2°C to 8°C Melting Point: 65-68° Beilstein Reference: 3202445 MDL No.: MFCD00014398

Formula: C₁₄H₁₄O₂
Melting Pt: 65…68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014398
CAS Number: 720-44-5
EINECS: 211-953-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR