tert-Nitrosobutane dimer ≥98%
Supplier: Thermo Scientific Chemicals
L00462-03
L00462-MD
CAAAAL00462-03EA
147.1
CAD
CAAAAL00462-03
CAAAAL00462-MD
tert-Nitrosobutane dimer ≥98%
tert-Nitrosobutane dimer
Purity: 98+%
CAS No.: 6841-96-9
Molecular Formula: C8H18N2O2
Formula Weight: 174.24
Storage Temperature: 2°C to 8°C
Melting Point: 88-92° subl.
MDL No.: MFCD00002065
Formula:
C₈H₁₈N₂O₂ Melting Pt: 88…92 °C (subl.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002065 CAS Number: 6841-96-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...