Ethyl (E)-3-(1-pyrrolidinyl)crotonate 97%

Supplier: Thermo Scientific Chemicals
Warning

L00461-06 L00461-14
CAAAAL00461-06EA 44.71 CAD
CAAAAL00461-06 CAAAAL00461-14 CAAAAL00461-22
Ethyl (E)-3-(1-pyrrolidinyl)crotonate 97%
Ethyl (E)-3-(1-pyrrolidinyl)crotonate

Purity: 97% CAS No.: 54716-02-8 Molecular Formula: C10H17NO2 Formula Weight: 183.25 Storage Temperature: 2°C to 8°C Refractive Index: 1.5410 Boiling Point: 125-126°/0.8mm Beilstein Reference: 1310113 MDL No.: MFCD00014097

Formula: C₁₀H₁₇NO₂
Boiling Pt: 125…126 °C/0.8 mm
Storage Temperature: Refrigerator
MDL Number: MFCD00014097
CAS Number: 54716-02-8
EINECS: 259-303-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR