2-Chloroacrylic acid 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L00350-04
L00350-09
CAAAAL00350-04EA
141.41
CAD
CAAAAL00350-04
CAAAAL00350-09
2-Chloroacrylic acid 96%
2-Chloroacrylic acid
Purity: 98+%
CAS No.: 598-79-8
Molecular Formula: C3H3ClO2
Formula Weight: 106.51
Storage Temperature: -30°C to -10°C
Melting Point: 63-67°
Beilstein Reference: 1098503
UN No.: 3261
MDL No.: MFCD00014336
Formula:
C₃H₃ClO₂ Melting Pt: 63…67 °C Storage Temperature: Freezer |
MDL Number:
MFCD00014336 CAS Number: 598-79-8 EINECS: 209-953-9 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...