1-(1-Cyclohexen-1-yl)piperidine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L00335-06
L00335-14
CAAAAL00335-06EA
79.16
CAD
CAAAAL00335-06
CAAAAL00335-14
1-(1-Cyclohexen-1-yl)piperidine 97%
1-(1-Cyclohexen-1-yl)piperidine
Purity: 97%
CAS No.: 2981-10-4
Molecular Formula: C11H19N
Formula Weight: 165.28
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5140
Boiling Point: 120-122°/16mm
Beilstein Reference: 113990
MDL No.: MFCD00014643
Formula:
C₁₁H₁₉N Boiling Pt: 120…122 °C/16 mm Storage Temperature: Refrigerator |
MDL Number:
MFCD00014643 CAS Number: 2981-10-4 UN: 3267 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...