4-Nitrophenyl formate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00316-06
L00316-14
CAAAAL00316-06EA
93.38
CAD
CAAAAL00316-06
CAAAAL00316-14
4-Nitrophenyl formate 98%
4-Nitrophenyl formate
Fieser: 1,744 21,285
Purity: 98%
CAS No.: 1865-01-6
Molecular Formula: C7H5NO4
Formula Weight: 167.12
Storage Temperature: 2°C to 8°C
Melting Point: 70-74°
Beilstein Reference: 1950897
MDL No.: MFCD00014713
Formula:
C₇H₅NO₄ Melting Pt: 70…74 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00014713 CAS Number: 1865-01-6 EINECS: 217-473-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...